missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrakis(pentafluorophenyl)borate, 97%

Product Code. p-7032985
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15417667

Brand: Thermo Scientific Alfa Aesar H36527.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89171-23-3 | C24BF20K | 718.14 g/mol

Potassium tetrakis(pentafluorophenyl)borate is used as a biochemical reagent, a catalyst and a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium tetrakis(pentafluorophenyl)borate is used as a biochemical reagent, a catalyst and a pharmaceutical intermediate.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89171-23-3
Molecular Formula C24BF20K
Molecular Weight (g/mol) 718.14
MDL Number MFCD06797410
InChI Key GYBHRIJOPWTIKA-UHFFFAOYSA-N
Synonym potassium tetrakis pentafluorophenyl borate, potassium tetrakis pentafluorophe-nyl borate, tetrakis-pentafluorophenyl-potassium borate, potassium tetrakis 2,3,4,5,6-pentafluorophenyl boranuide
PubChem CID 23693577
IUPAC Name potassium;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
SMILES [B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.[K+]

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 718.13
Percent Purity 97%
Chemical Name or Material Potassium tetrakis(pentafluorophenyl)borate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.