missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium tetracyanoplatinate(II) trihydrate, 99.9% (metals basis), Pt 44.9% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Potassium tetracyanoplatinate(II) trihydrate is used in the preparation other salts like Krogmann's salt, that are highly conducting, one dimensional materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Crystalline |
Quantity | 1 g |
Linear Formula | K2[Pt(CN)4]·3H2O |
UN Number | UN1588 |
Merck Index | 14,7685 |
Solubility Information | Soluble in hot water |
Formula Weight | 431.42 (377.37 Anhydrous) |
Percent Purity | 99.9% |
Concentration or Composition (by Analyte or Components) | Pt 44.9% min. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'