missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetracyanoplatinate(II) trihydrate, 99.9% (metals basis), Pt 44.9% min

Product Code. 11366048
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366048 5 g 5g
11376048 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11366048 Supplier Thermo Scientific Alfa Aesar Supplier No. 010536.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 14323-36-5 | C4H6K2N4O3Pt | 431.40 g/mol

Potassium tetracyanoplatinate(II) trihydrate is used in the preparation other salts like Krogmann's salt, that are highly conducting, one dimensional materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14323-36-5
Molecular Formula C4H6K2N4O3Pt
Molecular Weight (g/mol) 431.40
MDL Number MFCD00011374
InChI Key KBALGZKVNFJSRU-UHFFFAOYSA-N
IUPAC Name dipotassium tetracyanoplatinumbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].N#C[Pt++](C#N)(C#N)C#N

Specifications

Color White
Physical Form Crystalline
Quantity 5 g
Linear Formula K2[Pt(CN)4]·3H2O
UN Number UN1588
Merck Index 14,7685
Solubility Information Soluble in hot water
Formula Weight 431.42 (377.37 Anhydrous)
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pt 44.9% min.
Odor Odorless
Assay (metals basis)
Chemical Name or Material Potassium tetracyanoplatinate(II) trihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.