missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium tetrachloroplatinate(II), Premion™, 99.99% (metals basis), Pt 46.4% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Potassium tetrachloroplatinate(II) is used as important reagent for the preparation of other coordination complexes of platinum. It is also used in the induction of apoptosis in human breast cancer cells. Also used in the preparation of camptothecin-linked platinum anticancer agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | (decomposition) |
Color | Red to Brown |
Physical Form | Crystalline |
Quantity | 1 g |
Linear Formula | K2PtCl4 |
UN Number | UN3288 |
Merck Index | 14,7682 |
Solubility Information | Soluble in water (9.3g/L). Insoluble in ethyl alcohol. |
IUPAC Name | dipotassium tetrachloroplatinumdiuide |
Formula Weight | 415.11 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'