missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrachloroplatinate(II), Premion™, 99.99% (metals basis), Pt 46.4% min

Product Code. 11304998
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11304998

Brand: Thermo Scientific Alfa Aesar 043946.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10025-99-7 | Cl4H4K2Pt | 419.11 g/mol

Potassium tetrachloroplatinate(II) is used as important reagent for the preparation of other coordination complexes of platinum. It is also used in the induction of apoptosis in human breast cancer cells. Also used in the preparation of camptothecin-linked platinum anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-99-7
Molecular Formula Cl4H4K2Pt
Molecular Weight (g/mol) 419.11
MDL Number MFCD00011378
InChI Key JCAXKWJYKUEOFO-UHFFFAOYSA-N
Synonym potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate
PubChem CID 61440
SMILES Cl.Cl.Cl.Cl.[K+].[K+].[Pt--]

Specifications

Melting Point (decomposition)
Color Red to Brown
Physical Form Crystalline
Quantity 5 g
Linear Formula K2PtCl4
UN Number UN3288
Merck Index 14,7682
Solubility Information Soluble in water (9.3g/L). Insoluble in ethyl alcohol.
IUPAC Name dipotassium;tetrachloroplatinum(2-)
Formula Weight 415.11
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Pt 46.4% min
Odor Odorless
Density 3.38 g/mL
Assay (metals basis)
Chemical Name or Material Potassium tetrachloroplatinate (II)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.