missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrachloroplatinate(II), 99.9% (metals basis), Pt 46.0% min

Product Code. 11391688
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391688 5 g 5g
11301698 1 g 1g
11381688 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11391688 Supplier Thermo Scientific Alfa Aesar Supplier No. 011048.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 10025-99-7 | Cl4H4K2Pt | 419.11 g/mol

Potassium tetrachloroplatinate solution is used in the preparation of platinum (Pt) nanoparticles. Ligands like ammonia or triphenyl phosphine can able to replace the chloride ligands to afford various derivatives. The replacement by ammonia results in the preparation of cisplatin, which is used in the cancer treatment. Further, it is used as catalyst in the hydroarylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-99-7
Molecular Formula Cl4H4K2Pt
Molecular Weight (g/mol) 419.11
MDL Number MFCD00011378
InChI Key JCAXKWJYKUEOFO-UHFFFAOYSA-N
Synonym potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate
PubChem CID 61440
SMILES Cl.Cl.Cl.Cl.[K+].[K+].[Pt--]

Specifications

Melting Point (decomposition)
Color Red to Brown
Physical Form Crystalline
Quantity 5 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Soluble in hot water. Slightly soluble in water.
IUPAC Name dipotassium;tetrachloroplatinum(2-)
Formula Weight 415.11
Odor Odorless
Appearance Red-brown crystals
Density 3.38
Chemical Name or Material Potassium tetrachloroplatinate(II), Pt 46.0% min.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.