missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrachloroplatinate, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Catalog Number 11386315
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
10025-99-7
Cl4H4K2Pt
419.11
MFCD00011378
JCAXKWJYKUEOFO-UHFFFAOYSA-N
potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate
61440
dipotassium tetrachloroplatinumdiuide
Cl.Cl.Cl.Cl.[K+].[K+].[Pt--]
10025-99-7
Cl4H4K2Pt
419.11
MFCD00011378
JCAXKWJYKUEOFO-UHFFFAOYSA-N
potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate
61440
dipotassium tetrachloroplatinumdiuide
Cl.Cl.Cl.Cl.[K+].[K+].[Pt--]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY
Melting Point 250.0°C
Color Brown or Orange to Red
Physical Form Crystals
Quantity 1 g
Assay Percent Range 99.99%
Linear Formula K2PtCl4
Merck Index 15,7801
Solubility Information Solubility in water: 10g/L (20°C)
Formula Weight 415.1
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Potassium tetrachloroplatinate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'