missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrachloropalladate(II), Premion™, 99.99% (metals basis), Pd 32.2% min

Product Code. p-4824901
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11301978

Brand: Thermo Scientific Alfa Aesar 011885.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10025-98-6 | Cl4K2Pd | 326.417 g/mol

Potassium tetrachloropalladate(II) is widely used as a reagent and as a source of palladium chloride to make other palladium complexes and palladium nanoparticles. It finds an application in the synthesis of semiconducting metal-containing polymers. It also reacts with bis(dithiolates) to form metal-bis(dithiolates), which is used in bar code material, laser Q-switch materials, optical CD recording media and superconductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-98-6
Molecular Formula Cl4K2Pd
Molecular Weight (g/mol) 326.417
MDL Number MFCD00011373
InChI Key LGCKLDWLSVFMGL-UHFFFAOYSA-J
Synonym potassium tetrachloropalladate ii, potassium chloropalladite, potassium tetrachloropalladate, ccris 6625, potassium chlorophalladite, potassium palladium chloride, potassium palladous chloride, dipotassium tetrachloropalladate ii, dipotassium tetrachloropalladate 2-, tetrachloropalladate 2-dipotassium
PubChem CID 61438
IUPAC Name dipotassium;tetrachloropalladium(2-)
SMILES Cl[Pd-2](Cl)(Cl)Cl.[K+].[K+]

Specifications

Melting Point 105°C (decomposition)
Color Red to Brown
Physical Form Crystalline
Quantity 2 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water,methanol and ethanol.
Formula Weight 326.42
Odor Odorless
Sensitivity Hygroscopic
Appearance Red-brown crystals
Density 2.67
Chemical Name or Material Potassium tetrachloropalladate(II), Pd 32.2% min.
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.