missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrachloropalladate(II), Premion™, 99.99% (metals basis), Pd 32.2% min

Product Code. 11311978
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11311978 10 g 10g
11301978 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311978 Supplier Thermo Scientific Alfa Aesar Supplier No. 011885.09
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10025-98-6 | Cl4K2Pd | 326.417 g/mol

Potassium tetrachloropalladate(II) is widely used as a reagent and as a source of palladium chloride to make other palladium complexes and palladium nanoparticles. It finds an application in the synthesis of semiconducting metal-containing polymers. It also reacts with bis(dithiolates) to form metal-bis(dithiolates), which is used in bar code material, laser Q-switch materials, optical CD recording media and superconductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-98-6
Molecular Formula Cl4K2Pd
Molecular Weight (g/mol) 326.417
MDL Number MFCD00011373
InChI Key LGCKLDWLSVFMGL-UHFFFAOYSA-J
Synonym potassium tetrachloropalladate ii, potassium chloropalladite, potassium tetrachloropalladate, ccris 6625, potassium chlorophalladite, potassium palladium chloride, potassium palladous chloride, dipotassium tetrachloropalladate ii, dipotassium tetrachloropalladate 2-, tetrachloropalladate 2-dipotassium
PubChem CID 61438
IUPAC Name dipotassium;tetrachloropalladium(2-)
SMILES Cl[Pd-2](Cl)(Cl)Cl.[K+].[K+]

Specifications

Melting Point 105°C (decomposition)
Color Red to Brown
Physical Form Crystalline
Quantity 10 g
Odor Odorless
Sensitivity Hygroscopic
Appearance Red-brown crystals
Assay Percent Range 99.99% (metals basis)
Density 2.67
Solubility Information Soluble in water,methanol and ethanol.
Formula Weight 326.42
Chemical Name or Material Potassium tetrachloropalladate(II), Pd 32.2% min.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.