missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrabromoplatinate(II), 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 15407005
Click to view available options
Quantity:
500 mg
2.5 g
This item is not returnable. View return policy
13826-94-3
Br4K2Pt
592.90
MFCD00011376
AXZAYXJCENRGIM-UHFFFAOYSA-J
br4pt.2k, dipotassium;tetrabromoplatinum 2-, dipotassium tetrabromoplatinumdiuide
71310129
dipotassium;tetrabromoplatinum(2-)
[K+].[K+].[Br-].[Br-].[Br-].[Br-].[Pt++]
This item is not returnable. View return policy
13826-94-3
Br4K2Pt
592.90
MFCD00011376
AXZAYXJCENRGIM-UHFFFAOYSA-J
br4pt.2k, dipotassium;tetrabromoplatinum 2-, dipotassium tetrabromoplatinumdiuide
71310129
dipotassium;tetrabromoplatinum(2-)
[K+].[K+].[Br-].[Br-].[Br-].[Br-].[Pt++]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Physical Form Crystalline
Quantity 500 mg
Linear Formula K2PtBr4
Formula Weight 592.9
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Potassium tetrabromoplatinate (II)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'