missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrabromoaurate(III) dihydrate, Premion™, 99.99% (metals basis), Au 33% min

Product Code. 11347497
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11347497

Brand: Thermo Scientific Alfa Aesar 012664.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13005-38-4 | KAuBr4·2H2O

It is used as a Laboratory chemical and manufacture of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13005-38-4
Molecular Formula KAuBr4·2H2O
MDL Number MFCD00049650
Synonym Gold potassium bromide

Specifications

Color Red to Brown
Physical Form Crystalline
Quantity 1 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water and alcohol
Formula Weight 591.72 (555.69 Anhydrous)
Odor Weak
Appearance Red-brown crystals
Chemical Name or Material Potassium tetrabromoaurate(III) dihydrate, Au 33% min.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.