missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetraborate tetrahydrate, 99%

Product Code. 11398218
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398218 2 kg 2kg
11308228 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398218 Supplier Thermo Scientific Alfa Aesar Supplier No. 039435.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 12045-78-2 | K2B4O7+4H2O

It is an excellent solvent for metallic oxides at high temperatures. It is a good buffering agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12045-78-2
Molecular Formula K2B4O7+4H2O
MDL Number MFCD00149924
Synonym Potassium borate (tetra)

Specifications

Melting Point (decomposition)
Color White
Physical Form Granules
Quantity 2 kg
Assay Percent Range 99%
Solubility Information Soluble in water
Formula Weight 305.51 (233.44 Anhydrous)
Odor Odorless
Appearance White crystals
Density 1.74 (anhy)
Chemical Name or Material Potassium tetraborate tetrahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.