missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Tetraborate Tetrahydrate, ≥99.0% (T), Honeywell™
Shop All Honeywell Research Chemicals ProductsDescription
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Specifications
| Melting Point | >500°C |
| CAS | 12045-78-2 |
| Color | White |
| Physical Form | Crystalline |
| Quantity | 1 kg |
| Linear Formula | K2B4O7 · 4H2O |
| MDL Number | MFCD00149924 |
| UN Number | NONH for all modes of transport |
| Formula Weight | 305.5g/mol |
| Percent Purity | ≥99.0% (T) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?