missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-pentyloxide, 25% w/w in toluene

Product Code. 11398136
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398136 250 mL 250mL
11308146 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398136 Supplier Thermo Scientific Alfa Aesar Supplier No. L15111.AK
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 41233-93-6 | C5H11KO | 126.24 g/mol

Potassium tert-pentyloxide is a strong, hindered base used in combination with lithium diisopropyl amide to generate a superbase for the selective metallation of the aralkyl system at the 6-position of protected estradiols. It is used in organic synthesis, agrochemicals, pharmaceuticals, colorants and aroma chemicals. It acts as a catalyst in polymerization and isomerizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium tert-pentyloxide is a strong, hindered base used in combination with lithium diisopropyl amide to generate a superbase for the selective metallation of the aralkyl system at the 6-position of protected estradiols. It is used in organic synthesis, agrochemicals, pharmaceuticals, colorants and aroma chemicals. It acts as a catalyst in polymerization and isomerizations.

Notes
Air and moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and acids. Reacts violently with water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41233-93-6
Molecular Formula C5H11KO
Molecular Weight (g/mol) 126.24
MDL Number MFCD00064808
InChI Key ZRLVQFQTCMUIRM-UHFFFAOYSA-N
Synonym potassium 2-methylbutan-2-olate, potassium tert-amylate, potassium tert-pentoxide, potassium tert-pentylate, potassium tert-pentoxide solution, potassium 2-methyl-2-butoxide, 2-butanol, 2-methyl-, potassium salt, potassium tert-amyloxide, 2-butanol, 2-methyl-, potassium salt 1:1
PubChem CID 23683543
IUPAC Name potassium;2-methylbutan-2-olate
SMILES [K+].CCC(C)(C)[O-]

Specifications

Density 0.87
Flash Point 4°C (39°F)
Quantity 250 mL
UN Number UN2924
Beilstein 4149025
Sensitivity Air and moisture sensitive
Formula Weight 126.24
Concentration or Composition (by Analyte or Components) 25% w/w in toluene
Chemical Name or Material Potassium tert-pentyloxide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium tert-pentyloxide, 25% w/w in toluene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Toluene, Potassium tert-pentyloxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.