missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-butoxide, 97%

Product Code. p-7021560
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1000 g
Unit Size:
1000g
100g
250g
25g
500g
This item is not returnable. View return policy

Product Code. 11494010

Brand: Thermo Scientific Alfa Aesar A13947.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 865-47-4 | C4H9KO | 112.213 g/mol

Potassium tert-butoxide is used as a strong non-nucleophilic base in organic chemistry. It plays an active role in dehydrohalogenation reactions. It is also useful for greener amidation of esters. It serves as an intermediate in Mizoroki-Heck-type reactions. Furthermore, it is used as an initiator in anionic polymerization of carbazolyl-substituted oxiranes. It catalyzes the reaction of hydrosilanes and heterocyclic compounds to give the silyl derivatives with evolution of hydrogen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium tert-butoxide is used as a strong non-nucleophilic base in organic chemistry. It plays an active role in dehydrohalogenation reactions. It is also useful for greener amidation of esters. It serves as an intermediate in Mizoroki-Heck-type reactions. Furthermore, it is used as an initiator in anionic polymerization of carbazolyl-substituted oxiranes. It catalyzes the reaction of hydrosilanes and heterocyclic compounds to give the silyl derivatives with evolution of hydrogen.

Solubility
Soluble in hexane, toluene, diethyl ether and terahydrofuran.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water, acids, reducing agents, oxygen, alcohols, chlorinated solvents, halogens and ketones.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-47-4
Molecular Formula C4H9KO
Molecular Weight (g/mol) 112.213
MDL Number MFCD00012162
InChI Key LPNYRYFBWFDTMA-UHFFFAOYSA-N
Synonym potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
PubChem CID 23665647
IUPAC Name potassium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].[K+]

Specifications

Melting Point ∼255°C (decomposition)
Linear Formula (CH3)3COK
Quantity 25 g
UN Number UN3206
Beilstein 3556712
Sensitivity Air and moisture sensitive
Solubility Information Soluble in hexane,toluene,diethyl ether and terahydrofuran.
Formula Weight 112.22
Percent Purity 97%
Chemical Name or Material Potassium tert-butoxide
compliance-icons compliance-icons
Product Identifier
  • Potassium tert-butoxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 1
  • Self-heating substance Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H228-Flammable solid.
  • H252-Self-heating in large quantities; may catch fire.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Potassium tert-butoxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.