missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-butoxide, 2M (25% w/w) solution in 2-MeTHF, AcroSeal™

Product Code. p-4440155 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy

Product Code. 12675007

Brand: Thermo Scientific Acros 445831000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 865-47-4 | C4H9KO | 112.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium tert-butoxide
CAS 865-47-4, 96-47-9
Quantity 100 mL
Molecular Formula C4H9KO
Linear Formula (CH3)3COK
Flash Point -11°C
Synonym potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
Solubility Information Solubility in water: reacts
InChI Key LPNYRYFBWFDTMA-UHFFFAOYSA-N
SMILES CC(C)(C)[O-].[K+]
IUPAC Name potassium;2-methylpropan-2-olate
Molecular Weight (g/mol) 112.21
PubChem CID 23665647
Formula Weight 112.21
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.86
Density 0.8600g/mL
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium tert-butoxide, 2M (25% w/w) solution in 2-MeTHF
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402-Store in a dry place.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.