missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sulphite, MP Biomedicals

Product Code. 11423426 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423426 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11423426 Supplier MP Biomedicals Supplier No. 0522585525
 Restricted Item
Estimated Shipment: 19-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 10117-38-1
Quantity 25 g
Molecular Formula K2O3S
Synonym potassium sulfite, dipotassium sulfite, potassium sulphite, sulfurous acid, dipotassium salt, unii-015kzc652e, sulfurous acid, potassium salt 1:2, potassium sulfite k2so3, kaliumsulfit, potassiumsulfite, dipotassium ion sulfite
InChI Key BHZRJJOHZFYXTO-UHFFFAOYSA-L
SMILES [O-]S(=O)[O-].[K+].[K+]
IUPAC Name dipotassium;sulfite
Molecular Weight (g/mol) 158.254
PubChem CID 24958
Formula Weight 158.26
Chemical Name or Material Potassium Sulphite
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.