missing translation for 'onlineSavingsMsg'
Learn More

Potassium sulfate, meets analytical spec. of Ph. Eur., 99-101%, Solstice
BIOPHARMA

Product Code. 15671180
Change view
Click to view available options
Quantity:
1 kg
50 kg
6 x 1 kg
Packaging:
Fiberboard Box
Poly bottle
Unit Size:
1kg
50kg
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15671180 50 kg Fiberboard Box 50kg
15661180 1 kg Poly bottle 1kg
15681180 6 x 1 kg Poly bottle Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15671180 Supplier Honeywell-Fluka Supplier No. 1265850KG
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Meets analytical specification of Ph. Eur., 99-101%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1,698°C
CAS Max % 100.0000%
Physical Form Solid
Quantity 50 kg
Assay Percent Range 99 to 101%
Linear Formula K2SO4
IUPAC Name dipotassium;sulfate
Formula Weight 174.26
Percent Purity 99.0% to 101.0%
Grade Reagent Ph. Eur.
Packaging Fiberboard Box
Density 2.66 g/cm3
Chemical Name or Material Potassium sulfate, Meets analytical specification of Ph. Eur., 99-101%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.