missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sulfate, 99%

Product Code. 11461728
Change view
Click to view available options
Quantity:
1000 g
5000 g
Unit Size:
1000g
5000g
Product Code. Quantity unitSize
11461728 5000 g 5000g
11451728 1000 g 1000g
2 options
This item is not returnable. View return policy

Product Code. 11461728

Brand: Thermo Scientific Alfa Aesar A13975.0I

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7778-80-5 | K2O4S | 174.25 g/mol

Potassium sulfate is used in the production of fertilizers (i.e. providing potassium and sulfur), potassium alum, potassium carbonate and glass. In analytical chemistry, it is used in the determination of nitrogen by the Kjeldahl method and protein in foodstuffs. Also, it is used in the electrostatic effects on pKa values of amino acid residues in the staphylococcal nuclease. It is used in the separation of protein through electrophoresis and also serves as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium sulfate is used in the production of fertilizers (i.e. providing potassium and sulfur), potassium alum, potassium carbonate and glass. In analytical chemistry, it is used in the determination of nitrogen by the Kjeldahl method and protein in foodstuffs. Also, it is used in the electrostatic effects on pKa values of amino acid residues in the staphylococcal nuclease. It is used in the separation of protein through electrophoresis and also serves as a laboratory reagent.

Solubility
Soluble in water and glycerol. Insoluble in alcohol, acetate and carbon disulfide.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
IUPAC Name dipotassium sulfate
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1689°C
Melting Point 1,069°C
Quantity 5000 g
Merck Index 14,7674
Solubility Information Soluble in water and glycerol. Insoluble in alcohol,acetate and carbon disulfide.
Formula Weight 174.27
Percent Purity 99%
Odor Odorless
Density 2.662
Chemical Name or Material Potassium Sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.