missing translation for 'onlineSavingsMsg'
Learn More

Potassium sulfamate, 98%

Product Code. 11405907
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405907 500 g 500g
11485897 25 g 25g
11495897 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11405907 Supplier Thermo Scientific Alfa Aesar Supplier No. B24447.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13823-50-2 | H2KNO3S | 135.178 g/mol

Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13823-50-2
Molecular Formula H2KNO3S
Molecular Weight (g/mol) 135.178
MDL Number MFCD00059941
InChI Key BTAAXEFROUUDIL-UHFFFAOYSA-M
Synonym potassium sulfamate, potassium sulphamate, sulfamic acid,potassium salt 1:1, sulfamic acid, monopotassium salt, sulfamic acid, potassium salt 1:1, amidosulfonic acid, potassium salt, potassium amidosulfonate, acmc-209chf, sulfamic acid potassium salt, aminosulfonic acid potassium salt
PubChem CID 23687442
IUPAC Name potassium;sulfamate
SMILES NS(=O)(=O)[O-].[K+]

Specifications

Color White
Physical Form Crystalline Solid
Quantity 500 g
Linear Formula H2NSO3K
Formula Weight 135.18
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material Potassium sulfamate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.