missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium sulfamate, 98%, Thermo Scientific Chemicals
40.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications
Color | White |
Physical Form | Crystalline Solid |
Quantity | 100 g |
Linear Formula | H2NSO3K |
Formula Weight | 135.18 |
Percent Purity | 98% |
Sensitivity | Hygroscopic |
Chemical Name or Material | Potassium sulfamate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'