missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Standard Solution 1000ppm (1mg/mL), for Ion Pair Chromatography, Ready To Use, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Specifications
Specifications
| Packaging | HDPE plastic bottle |
| Vapor Pressure | 14mmHg at 20°C |
| Vapor Density | 0.7 |
| Physical Form | Liquid |
| MDL Number | 133776 |
| Color | Colorless |
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Quantity | 100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction