missing translation for 'onlineSavingsMsg'
Learn More

Potassium Standard for ICP/MS, 2% HCl, SPEX CertiPrep™

Product Code. 15255936 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15255936 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15255936 Supplier SPEX Certiprep Supplier No. CLK11BY

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7440-09-7, 7732-18-5
CAS Max % 2.0, 0.0, 98.0
Density 1.003 g/cm3 at 20°C
Physical Form Liquid
Molecular Formula K
MDL Number MFCD00133776
InChI Key ZLMJMSJWJFRBEC-UHFFFAOYSA-N
SMILES [K]
IUPAC Name potassium
Solvent or Matrix 2% HCl
Molecular Weight (g/mol) 39.10
PubChem CID 5462222
Quantity 125 mL
ChEBI CHEBI:26216
Grade Claritas PPT
Concentration or Composition (by Analyte or Components) 1mg/L
Chemical Name or Material Potassium Standard, ICP/MS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.