missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
£144.00
Chemical Identifiers
CAS | 7447-40-7, 7647-01-0, 7732-18-5 |
---|---|
Molecular Formula | ClK |
Molecular Weight (g/mol) | 74.55 |
MDL Number | MFCD00011360 |
InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
PubChem CID | 4873 |
ChEBI | CHEBI:32588 |
IUPAC Name | potassium;chloride |
SMILES | [Cl-].[K+] |
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
K | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7447-40-7, 7647-01-0, 7732-18-5 | |
74.55 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
CHEBI:32588 | |
[Cl-].[K+] |
ClK | |
MFCD00011360 | |
4873 | |
potassium;chloride |
Specifications
7447-40-7, 7647-01-0, 7732-18-5 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
ClK | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium;chloride | |
4873 | |
Assurance | |
Potassium Standard |
Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00011360 | |
[Cl-].[K+] | |
74.55 | |
CHEBI:32588 | |
2% HNO3 |
Safety and Handling
Causes severe skin burns and eye damage
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 1000
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : nitric acid
Product intended for laboratory use only