missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sorbate, TRC

Product Code. 30419395
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30419395 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419395 Supplier Toronto Research Chemicals Supplier No. TRCP698450100G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Sorbate
CAS 24634-61-5
Purity Grade Notes HPLC
Molecular Formula C6 H7 O2 . K
Quantity 100 g
Synonym Potassium Sorbate, (2E,4E)-2,4-Hexadienoic acid potassium salt (1:1), (2E,4E)-2,4-Hexadienoic acid potassium salt, (E,E)-2,4-Hexadienoic acid potassium salt, Sorbic acid potassium salt, E 202, E 202 (preservative), K 60/95, PO 300, Potassium (E,E)-hexa-2,4-dienoate, Potassium sorbate
InChI Formula InChI=1S/C6H8O2.K/c1-2-3-4-5-6(7)8;/h2-5H,1H3,(H,7,8);/q;+1/p-1/b3-2+,5-4+;
SMILES [K+].C\C=C\C=C\C(=O)[O-]
IUPAC Name potassium;(2E,4E)-hexa-2,4-dienoate
Molecular Weight (g/mol) 150.22
Formula Weight 150.0083
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.