missing translation for 'onlineSavingsMsg'
Learn More

Potassium sorbate, 99%

Product Code. 11405253
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405253 5000 g 5000g
11495243 250 g 250g
11485243 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11405253 Supplier Thermo Scientific Alfa Aesar Supplier No. A12844.0I
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24634-61-5 | C6H7KO2 | 150.218 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Potassium sorbate serves as an inhibitor for molds, yeasts in many food items like cheese, wine, yogurt, dried meats, apple cider, soft drinks, fruit drinks and baked goods. It is used in the production of syrup, milkshakes, creams, lotions, shampoos, makeup and sunscreen products. It is mainly used to increase the shelf life of various commercial products without affecting its taste, smell and color of the food. It is used as a preservative and considered to be a replacement for parabens.

Solubility
Soluble in water, alcohol, propylene glycol. Slightly soluble in acetoneand chloroform, corn oil and ether. Insoluble in benzene.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24634-61-5
Molecular Formula C6H7KO2
Molecular Weight (g/mol) 150.218
MDL Number MFCD00016546
InChI Key CHHHXKFHOYLYRE-STWYSWDKSA-M
Synonym potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c
PubChem CID 23676745
ChEBI CHEBI:77868
IUPAC Name potassium;(2E,4E)-hexa-2,4-dienoate
SMILES CC=CC=CC(=O)[O-].[K+]

Specifications

Melting Point >300°C
Density 1.361
pH 8.3
Assay Percent Range 99%
Linear Formula CH3CH=CH-CH=CHCO2K
Quantity 5000 g
Merck Index 14,7671
Solubility Information Soluble in water,alcohol,propylene glycol. Slightly soluble in acetoneand chloroform,corn oil and ether. Insoluble in benzene.
Formula Weight 150.22
Percent Purity 99%
Chemical Name or Material Potassium sorbate
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.