missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sodium Tartrate Tetrahydrate, 99.0-102.0%, (ACS Reagent Grade), MP Biomedicals

Product Code. 11437852 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
3 kg
Unit Size:
100g
3kg
500g
This item is not returnable. View return policy

Product Code. 11437852

Brand: MP Biomedicals 0215256390

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6381-59-5
Molecular Formula C4H12KNaO10
Molecular Weight (g/mol) 282.218
InChI Key VZOPRCCTKLAGPN-UHFFFAOYSA-L
Synonym potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
PubChem CID 2724148
IUPAC Name potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
SMILES C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]

Specifications

Quantity 500 g
Assay Percent Range 99.0-102.0%
Formula Weight 282.22
Grade ACS Reagent
Chemical Name or Material Potassium Sodium Tartrate Tetrahydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.