missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sodium Tartrate Tetrahydrate, 99.0-102.0%, (ACS Reagent Grade), MP Biomedicals

Product Code. 11427852 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
Unit Size:
100g
3kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427852 100 g 100g
11437852 500 g 500g
11447852 3 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427852 Supplier MP Biomedicals Supplier No. 0215256380
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 6381-59-5
Molecular Formula C4H12KNaO10
Molecular Weight (g/mol) 282.218
InChI Key VZOPRCCTKLAGPN-UHFFFAOYSA-L
Synonym potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
PubChem CID 2724148
IUPAC Name potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
SMILES C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]

Specifications

Quantity 100 g
Assay Percent Range 99.0-102.0%
Formula Weight 282.22
Grade ACS Reagent
Chemical Name or Material Potassium Sodium Tartrate Tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.