missing translation for 'onlineSavingsMsg'
Learn More

Potassium silver cyanide, 99.9% (metals basis), Ag 54.2%

Product Code. 11356449
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11356449

Brand: Thermo Scientific Alfa Aesar 012551.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Silver plating | CAS: 506-61-6

Silver platingPotassium silver cyanide is used in silver plating and in the manufacturing of antiseptics. It is a versatile bridging ligand and finds use for multidimensional polymer construction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-61-6
MDL Number MFCD00036297
Synonym Potassium dicyanoargentate; Silver(I) potassium cyanide

Specifications

Color White
Physical Form Powder
Quantity 500 g
Linear Formula KAg(CN)2
UN Number UN1588
Merck Index 14,7669
Solubility Information Soluble in waterSoluble in water; Insoluble in acids.
Formula Weight 199.01
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Ag 54.2%
Sensitivity Light sensitive
Density 2.36 g/mL
Assay (metals basis)
Chemical Name or Material Potassium silver cyanide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.