missing translation for 'onlineSavingsMsg'
Learn More

Potassium silicate, anhydrous

Product Code. 11410850
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410850 500 g 500g
11400850 100 g 100g
11420850 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410850 Supplier Thermo Scientific Alfa Aesar Supplier No. 044493.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1312-76-1 | K2O3Si | 154.28 g/mol

Potassium silicate is used in binders, carbon electrodes, lead pencils, protective coatings, insoluble pigments, phosphors on television tubes and detergent additives. It acts as a catalyst to make glass, ceramics, welding rods, adhesives, cements, gels, paints, pulp and paper production. Further, it is used in horticulture as a soluble source of potassium and silicon. It acts as a corrosion inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-76-1
Molecular Formula K2O3Si
Molecular Weight (g/mol) 154.28
MDL Number MFCD00145152
InChI Key NNHHDJVEYQHLHG-UHFFFAOYSA-N
IUPAC Name dipotassium oxosilanebis(olate)
SMILES [K+].[K+].[O-][Si]([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 500 g
Solubility Information Soluble in water.
Odor Odorless
Chemical Name or Material Potassium silicate
compliance-icons
Product Identifier
  • Potassium silicate, anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Potassium silicate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.