missing translation for 'onlineSavingsMsg'
Learn More

Potassium selenite

Product Code. p-7050772
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11306499

Brand: Thermo Scientific Alfa Aesar 039436.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10431-47-7 | K2O3Se | 205.165 g/mol

One among the compounds used in cattle feeds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10431-47-7
Molecular Formula K2O3Se
Molecular Weight (g/mol) 205.165
MDL Number MFCD00049663
InChI Key RNGFNLJMTFPHBS-UHFFFAOYSA-L
Synonym potassium selenite, dipotassium selenite, ccris 6885, selenious acid, dipotassium salt, kaliumselenat, acmc-20ajua, selenious acid, potassium salt 1:2, dipotassium ion selenit, selenious acid,potassium salt 1:2
PubChem CID 150192
IUPAC Name dipotassium;selenite
SMILES [O-][Se](=O)[O-].[K+].[K+]

Specifications

Melting Point 875°C (decomposition)
Color White
Physical Form Powder
Quantity 25 g
UN Number UN2630
Solubility Information Soluble in ethyl alcohol,water.
Formula Weight 205.16
Sensitivity Hygroscopic
Density 2.851 g/mL
Chemical Name or Material Potassium selenite

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.