missing translation for 'onlineSavingsMsg'
Learn More

Potassium selenite

Product Code. 11306499
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11306499 25 g 25g
11316499 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11306499 Supplier Thermo Scientific Alfa Aesar Supplier No. 039436.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10431-47-7 | K2O3Se | 205.165 g/mol

One among the compounds used in cattle feeds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10431-47-7
Molecular Formula K2O3Se
Molecular Weight (g/mol) 205.165
MDL Number MFCD00049663
InChI Key RNGFNLJMTFPHBS-UHFFFAOYSA-L
Synonym potassium selenite, dipotassium selenite, ccris 6885, selenious acid, dipotassium salt, kaliumselenat, acmc-20ajua, selenious acid, potassium salt 1:2, dipotassium ion selenit, selenious acid,potassium salt 1:2
PubChem CID 150192
IUPAC Name dipotassium;selenite
SMILES [O-][Se](=O)[O-].[K+].[K+]

Specifications

Melting Point 875°C (decomposition)
Color White
Physical Form Powder
Quantity 25 g
UN Number UN2630
Solubility Information Soluble in ethyl alcohol,water.
Formula Weight 205.16
Sensitivity Hygroscopic
Density 2.851 g/mL
Chemical Name or Material Potassium selenite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.