missing translation for 'onlineSavingsMsg'
Learn More

Potassium selenate, 99.5% (metals basis)

Product Code. 11440950
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440950 2 g 2g
11420950 10 g 10g
11430950 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440950 Supplier Thermo Scientific Alfa Aesar Supplier No. 068111.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7790-59-2 | K2O4Se | 221.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-59-2
Molecular Formula K2O4Se
Molecular Weight (g/mol) 221.164
MDL Number MFCD00054125
InChI Key YAZJAPBTUDGMKO-UHFFFAOYSA-L
Synonym potassium selenate, dipotassium selenate, unii-99798pu7mi, selenic acid, dipotassium salt, 2k.so4e, selenic acid solution, selenic acid, potassium salt 1:2, selenic acid solution, 40 wt. % in water 100g
PubChem CID 62680
IUPAC Name dipotassium;selenate
SMILES [O-][Se](=O)(=O)[O-].[K+].[K+]

Specifications

Color White
Physical Form Powder
Quantity 2 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Soluble in water
Formula Weight 221.16
Odor Odorless
Sensitivity Hygroscopic
Density 3.07 g/cm3 at 20°C
Chemical Name or Material Potassium selenate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.