missing translation for 'onlineSavingsMsg'
Learn More

Potassium polysulfide, pure

Product Code. 10711594 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
250g
This item is not returnable. View return policy

Product Code. 10711594

Brand: Thermo Scientific 389590010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 37199-66-9 | (S)nK2 | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37199-66-9
Molecular Formula (S)nK2
Molecular Weight (g/mol) NaN
MDL Number MFCD00148074
InChI Key ZOCLAPYLSUCOGI-UHFFFAOYSA-N
Synonym potassium sulfane, potassium polysulfide, caswell no. 701, potassium sulfide k2 sx, epa pesticide chemical code 076703
PubChem CID 5362529
SMILES [K+].[K+].*-[S--]-*

Specifications

Melting Point 200.0°C to 250.0°C
Quantity 1 kg
Linear Formula (K2S)n
Solubility Information Solubility in water: 500g/l (20°C)
IUPAC Name potassium sulfane
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Potassium polysulfide
compliance-icons compliance-icons
Product Identifier
  • Potassium polysulfide
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.