Learn More
Potassium phthalimide, 98+%, Thermo Scientific Chemicals
Description
Potassium phthalimide is used as an intermediate in the synthesis of N-alkylated phthalimides, which is involved in the preparation of primary amines (Gabriel synthesis) by the hydrolysis reaction. It is also used as an intermediate for synthetic indigo, pigments, dyes and pharmaceuticals. Further, it is employed as an organocatalyst for the cyanosilylation of various carbonyl compounds under extremely mild conditions. In addition to this, it serves as a reagent for the transformation of allyl- and alkyl halides into protected primary amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium phthalimide is used as an intermediate in the synthesis of N-alkylated phthalimides, which is involved in the preparation of primary amines (Gabriel synthesis) by the hydrolysis reaction. It is also used as an intermediate for synthetic indigo, pigments, dyes and pharmaceuticals. Further, it is employed as an organocatalyst for the cyanosilylation of various carbonyl compounds under extremely mild conditions. In addition to this, it serves as a reagent for the transformation of allyl- and alkyl halides into protected primary amines.
Solubility
Soluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong acids.
Specifications
Melting Point | >300°C |
Density | 1.63 |
Quantity | 500 g |
Beilstein | 3598719 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in water. |
Formula Weight | 185.23 |
Percent Purity | ≥98% |
Chemical Name or Material | Potassium phthalimide |
RUO – Research Use Only