missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phthalate Monobasic, ≥99.5% (T), For HPLC, Solstice

Product Code. 15607570
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly Bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15607570 250 g Poly Bottle 250g
15617570 50 g Poly Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15607570 Supplier Honeywell-Fluka Supplier No. 96148250G
 Restricted Item
Estimated Shipment: 20-04-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for HPLC,≥99.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
MDL Number MFCD00013070
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Boiling Point 378°C
Melting Point 295°C to 300°C
Color Colorless
pH 3.5 to 5
Physical Form Solid
Quantity 250 g
Loss on Drying max. 0.5% (105°C)
Odor Odorless
Packaging Poly Bottle
Density 1.64g/cm3
Linear Formula HOOCC6H4COOK
UN Number NONH for all modes of transport
Beilstein 3637128
Formula Weight 204.22g/mol
Percent Purity ≥99.5% (T)
Grade Not Available
Chemical Name or Material Potassium phthalate monobasic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.