missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium phosphate, tribasic monohydrate, 96%, extra pure
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1340.0°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 25 kg |
| Assay Percent Range | 96% |
| Linear Formula | K3PO4·H2O |
| Solubility Information | Solubility in water: soluble. Other solubilities: insoluble in alcohol |
| Formula Weight | 230.28 |
| Percent Purity | 96% |
| Grade | Extra Pure |
| Show More |
Safety and Handling
Product Identifier
- Potassium phosphate tribasic monohydrate
- Danger
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H335-May cause respiratory irritation.
- H318-Causes serious eye damage.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?