missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium phosphate, tribasic, 97%, pure, anhydrous, Thermo Scientific Chemicals
35.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1340.0°C |
Color | White |
Physical Form | Powder, Granules or Chunks |
Quantity | 500 g |
Assay Percent Range | 97% |
Linear Formula | K3PO4 |
Merck Index | 15,7781 |
Solubility Information | Solubility in water: 508g/L (25°C). Other solubilities: insoluble in alcohol |
Formula Weight | 212.27 |
Percent Purity | 97% |
Show More |
Safety and Handling
Product Identifier
- Potassium phosphate tribasic
- Danger
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'