missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium phosphate monohydrate, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2 kg
Description
Potassium phosphate monohydrate acts as a reagent and buffer used in molecular biology, biochemistry, and chromatography. It is utilized to study the effects of freezing and thawing of proteins sensitive to conformational changes. It is also used in the extraction of keratohyalin protein from bovine tissue. Further, it is used as a food additive, emulsifier, foaming agent and fertilizer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Granules |
Quantity | 2 kg |
Solubility Information | Soluble in water. Insoluble in alcohol. |
Formula Weight | 230.28 (212.27 Anhydrous) |
Odor | Odorless |
Appearance | White granules |
Density | 2.564 |
Chemical Name or Material | Potassium phosphate monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'