missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate, Monobasic, ULTREX™ Ultrapure Reagent, J.T. Baker™

Catalog Number p-5390855
646.00 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
100 g
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
7778-77-0
H2KO4P
136.08
MFCD00011401 MFCD00147253
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
516951
CHEBI:63036
[K+].OP(O)([O-])=O
This item is not returnable. View return policy
7778-77-0
H2KO4P
136.08
MFCD00011401 MFCD00147253
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
516951
CHEBI:63036
[K+].OP(O)([O-])=O
Quantity 100 g
IUPAC Name potassium;dihydrogen phosphate
Formula Weight 136.09
Grade ULTREX™ Ultrapure Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.34kg
Chemical Name or Material Potassium Phosphate, Monobasic
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'