missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate, Monobasic, ULTREX™ Ultrapure Reagent, J.T. Baker™
646.00 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
100 g
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
Quantity | 100 g |
IUPAC Name | potassium;dihydrogen phosphate |
Formula Weight | 136.09 |
Grade | ULTREX™ Ultrapure Reagent |
Packaging | Wide Mouth Amber Glass Bottle |
Specific Gravity | 1L = 2.34kg |
Chemical Name or Material | Potassium Phosphate, Monobasic |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'