missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Monobasic, MP Biomedicals™

Product Code. 11435382 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g

Product Code. 11435382

Brand: MP Biomedicals 02195453.1

Please to purchase this item. Need a web account? Register with us today!

Displays a high buffering capacity. Potassium Phosphate Monobasic, MP Biomedicals is a reagent that is used as a component for a wide variety of media used in the culture of microorganisms and in phosphate buffered saline (PBS). In addition to helping mai

It occurs in several forms: monobasic, dibasic and tribasic (K3PO4). Most pH neutral potassium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 100 g
Solubility Information Soluble in water; Insoluble to slightly soluble in ethanol; insoluble in alcohol.
Formula Weight 136.09
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.