missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate, monobasic, for HPLC

Product Code. 16554204 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
Product Code. Quantity unitSize
16554204 1 kg 1kg
16564204 50 g 50g
16574204 250 g 250g
3 options
This item is not returnable. View return policy

Product Code. 16554204

Brand: Thermo Scientific 447370010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7778-77-0 | H2KO4P | 136.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Melting Point 253.0°C
Quantity 1 kg
Linear Formula KH2PO4
Merck Index 15, 7780
Solubility Information Solubility aq. soln.: 222 g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 136.09
Grade HPLC
Chemical Name or Material Potassium phosphate, monobasic

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.