missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate monobasic, meets analytical spec. of NF, Ph. Eur., E340, Anhydrous, 98-100.5% (calc. to the dried substance), Honeywell Fluka™

Product Code. p-7071715 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
6 x 500 g
4 x 5 kg
50 kg
6 x 1 kg
Unit Size:
1kg
500g
50kg
5kg
Pack of 4
Pack of 6
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15690720 500 g 500g
15670720 1 kg 1kg
15630730 6 x 500 g Pack of 6
15610730 5 kg 5kg
15620730 6 x 1 kg Pack of 6
15680720 4 x 5 kg Pack of 4
15600730 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15690720 Supplier Honeywell-Fluka Supplier No. 04243500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Meets analytical specification of NF, Ph. Eur., E340, Anhydrous, 98-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
SMILES [K+].OP(O)([O-])=O

Specifications

Physical Form Anhydrous
Quantity 500 g
Linear Formula KH2PO4
UN Number NONH for all modes of transport
Solubility Information 4.5parts soluble in water, insoluble in alcohol
IUPAC Name potassium;dihydrogen phosphate
Formula Weight 136.09g/mol
Percent Purity 98 to 100.5% (calc. to the dried substance)
Packaging Plastic Bottle
Density 2.338g/mL at 25°C
Chemical Name or Material Potassium Phosphate Monobasic, Meets analytical specification of Ph. Eur.NF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.