missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate, Monobasic, Crystal, Multi-Compendial, N.F., J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
Unit Size:
2.5kg
500g
Specifications
Specifications
| Physical Form | Crystals |
| Quantity | 2.5 kg |
| IUPAC Name | potassium;dihydrogen phosphate |
| Formula Weight | 136.09 |
| Grade | Multi-Compendial/NF |
| Packaging | Wide Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 2.34kg |
| Chemical Name or Material | Potassium Phosphate, Monobasic |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction