missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Dibasic Trihydrate, (White, crystalline powder/Reagent Grade), MP Biomedicals

Product Code. 11440572 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440572 5 kg 5kg
11430572 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440572 Supplier MP Biomedicals Supplier No. 0219143205
 Restricted Item
Estimated Shipment: 22-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16788-57-1 | Formula: H7K2O7P | Weight: 228.22 g/mol

Chemical Identifiers

CAS 16788-57-1
Molecular Formula H7K2O7P
Molecular Weight (g/mol) 228.22
MDL Number MFCD00149926
InChI Key XQGPKZUNMMFTAL-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
PubChem CID 16217523
IUPAC Name dipotassium trihydrate hydrogen phosphate
SMILES O.O.O.[K+].[K+].OP([O-])([O-])=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 5 kg
Formula Weight 228.2
Percent Purity 99.8%
Grade Reagent
Chemical Name or Material Potassium Phosphate Dibasic Trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.