missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate dibasic trihydrate, Puriss. p.a., ≥99.0% (T), Solstice

Product Code. 16655873
Change view
Click to view available options
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16655873 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16655873 Supplier Fluka Supplier No. 6034925KG
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 16788-57-1
Color White
CAS Max % 1
Quantity 25 kg
Molecular Formula H7K2O7P
Linear Formula K2HPO4 · 3H2O
Synonym Dipotassium hydrogen phosphate trihydrate; Dipotassium phosphate; di-Potassium hydrogen phosphate trihydrate
InChI Key XQGPKZUNMMFTAL-UHFFFAOYSA-L
SMILES O.O.O.[K+].[K+].OP([O-])([O-])=O
IUPAC Name dipotassium trihydrate hydrogen phosphate
Molecular Weight (g/mol) 228.22
Grade Reagent
Chemical Name or Material Potassium phosphate dibasic trihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.