missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Dibasic Trihydrate, 99+%, (Crystalline), MP Biomedicals

Product Code. 11477202 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
10 kg
Unit Size:
10kg
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477202 1 kg 1kg
11487202 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11477202 Supplier MP Biomedicals Supplier No. 0215194601
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 16788-57-1
Molecular Formula H7K2O7P
Molecular Weight (g/mol) 228.22
MDL Number MFCD00149926
InChI Key XQGPKZUNMMFTAL-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
PubChem CID 16217523
IUPAC Name dipotassium trihydrate hydrogen phosphate
SMILES O.O.O.[K+].[K+].OP([O-])([O-])=O

Specifications

Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 99+%
Formula Weight 228.2
Chemical Name or Material Potassium Phosphate Dibasic Trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.