missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate, Dibasic, Powder, Ultrapure Bioreagent, J.T. Baker™

Catalog Number 15568204
309.70 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Liquid Chromatography and Molecular Biology applications
CAS 7758-11-4
Physical Form Powder
Quantity 500 g
Molecular Formula HK2O4P
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
SMILES OP(=O)([O-])[O-].[K+].[K+]
IUPAC Name dipotassium;hydrogen phosphate
Molecular Weight (g/mol) 174.17
PubChem CID 24450
ChEBI CHEBI:32031
Formula Weight 174.18
Grade Ultrapure Bioreagent
Packaging Poly Bottle
Chemical Name or Material Potassium Phosphate, Dibasic
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'