missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate, Dibasic, 1.0M solution in water

Product Code. 10173064 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10173064 1 L Glass bottle 1 L
10729005 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10173064 Supplier Thermo Scientific Acros Supplier No. 397080010
  • ONLINE EXCLUSIVE PRICE
    £34.65
    £33.60 / 1 L
    Save £1.05
    3% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7758-11-4 | HK2O4P | 174.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium phosphate, dibasic
CAS 7758-11-4, 7732-18-5
Quantity 1 L
Molecular Formula HK2O4P
Linear Formula K2HPO4
MDL Number MFCD00011383
Merck Index 15, 7779
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
SMILES OP(=O)([O-])[O-].[K+].[K+]
IUPAC Name dipotassium;hydrogen phosphate
Molecular Weight (g/mol) 174.18
PubChem CID 24450
ChEBI CHEBI:32031
Formula Weight 174.18
Concentration or Composition (by Analyte or Components) 1.0M, exact strength on the certificate of analysis
Packaging Glass bottle
Appearance Clear colorless solution
Specific Gravity 1.13
Density 1.1300g/mL
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.